siRNA Details
| Gene Symbol: Aig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025446.3 | NP_079722.1 | 3, 4 | 469 | ||
| GenBank mRNA | AK008210.1 | - | 3, 4 | 472 | ||
| AK162306.1 | - | 3, 4 | 488 | |||
| BC035305.1 | - | 3, 4 | 619 | 3980923 | ||
| EF410113.1 | - | 1, 2 | 354 | |||

Aig1
Genomic Map