siRNA Details
| Gene Symbol: Tm4sf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025453.3 | NP_079729.1 | 4 | 461 | ||
| GenBank mRNA | AK007532.1 | - | 4 | 462 | ||
| AK017154.1 | - | 4 | 463 | |||
| AK017195.1 | - | 4 | 462 | |||
| AK017209.1 | - | 4 | 470 | |||
| BC117736.2 | - | 4 | 453 | 40089849 | ||

Tm4sf20
Genomic Map