siRNA Details
| Gene Symbol: Eapp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025456.3 | NP_079732.1 | 4 | 479 | ||
| GenBank mRNA | AK007442.1 | - | 4 | 434 | ||
| AK012800.1 | - | 4 | 436 | |||
| AY882557.1 | - | 4 | 398 | |||
| BC037622.1 | - | 4 | 506 | 5373275 | ||

Eapp
Genomic Map