siRNA Details
| Gene Symbol: Eapp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025456.3 | NP_079732.1 | 4 | 445 | ||
| GenBank mRNA | AK007442.1 | - | 4 | 400 | ||
| AK012800.1 | - | 4 | 402 | |||
| AK145027.1 | - | 4 | 464 | |||
| AK145041.1 | - | 4 | 464 | |||
| AY882557.1 | - | 4 | 364 | |||
| BC037622.1 | - | 4 | 472 | 5373275 | ||

Eapp
Genomic Map