siRNA Details
| Gene Symbol: Senp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK010813.1 | - | 5 | 480 | ||
| AK012740.1 | - | 6 | 572 | |||
| AK013090.1 | - | 6 | 572 | |||
| AK020068.1 | - | 5 | 475 | |||
| AK083086.1 | - | 6 | 557 | |||
| AK085060.1 | - | 6 | 571 | |||
| AK122542.1 | - | 6 | 567 | |||
| AK164354.1 | - | 6 | 556 | |||
| BC058593.1 | - | 6 | 514 | 5364345 | ||
| CF583861.1 | - | 7 | 770 | |||

Senp7
Genomic Map