siRNA Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160208.1 | NP_001153680.1 | 5 | 818 | ||
| NM_001160209.1 | NP_001153681.1 | 4 | 905 | |||
| NM_025505.4 | NP_079781.2 | 6 | 839 | |||
| GenBank mRNA | AK005489.1 | - | 4 | 639 | ||
| AK006544.1 | - | 5 | 822 | |||
| AK029787.1 | - | 6 | 840 | |||
| AK167493.1 | - | 4 | 912 | |||
| AK168933.1 | - | 4 | 666 | |||
| AK171167.1 | - | 4 | 697 | |||
| BC027025.1 | - | 4 | 680 | 5374749 | ||

Blzf1
Genomic Map