siRNA Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160208.1 | NP_001153680.1 | 7 | 1113 | ||
| NM_001160209.1 | NP_001153681.1 | 6 | 1200 | |||
| NM_025505.4 | NP_079781.2 | 8 | 1134 | |||
| GenBank mRNA | AK005489.1 | - | 6 | 934 | ||
| AK006544.1 | - | 7 | 1117 | |||
| AK029787.1 | - | 8 | 1135 | |||
| AK167493.1 | - | 6 | 1207 | |||
| AK168933.1 | - | 6 | 961 | |||
| AK171167.1 | - | 6 | 992 | |||
| BC027025.1 | - | 6 | 975 | 5374749 | ||

Blzf1
Genomic Map