siRNA Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160208.1 | NP_001153680.1 | 4 | 459 | ||
| NM_001160209.1 | NP_001153681.1 | 3 | 546 | |||
| NM_025505.4 | NP_079781.2 | 5 | 480 | |||
| GenBank mRNA | AK005489.1 | - | 3 | 280 | ||
| AK006544.1 | - | 4 | 463 | |||
| AK029787.1 | - | 5 | 481 | |||
| AK167493.1 | - | 3 | 553 | |||
| AK168933.1 | - | 3 | 309 | |||
| AK171167.1 | - | 3 | 338 | |||
| BC027025.1 | - | 3 | 321 | 5374749 | ||

Blzf1
Genomic Map