siRNA Details
| Gene Symbol: Gmpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002326.1 | - | 2, 3 | 394 | ||
| AK009159.1 | - | 2, 3 | 405 | |||
| AK084508.1 | - | 2, 3 | 322 | |||
| AK179847.1 | - | 2, 3 | 145 | |||
| BC010827.1 | - | 2, 3 | 376 | 4219628 | ||

Gmpr
Genomic Map