siRNA Details
| Gene Symbol: 2310022A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122767.1 | NP_001116239.1 | 1, 2 | 139 | ||
| NM_175107.5 | NP_780316.3 | 3, 4 | 248 | |||
| GenBank mRNA | AK009456.1 | - | 1, 2 | 110 | ||
| AK009941.1 | - | 1, 2 | 139 | |||
| AK077369.1 | - | 1, 2 | 140 | |||
| AK140814.1 | - | 1, 2 | 119 | |||
| BB853451.1 | - | 3, 4 | 249 | |||
| BC023369.1 | - | 3, 4 | 227 | 5038597 | ||

2310022A10Rik
Genomic Map