siRNA Details
| Gene Symbol: Rtca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025517.3 | NP_079793.2 | 3 | 440 | ||
| GenBank mRNA | AK002543.1 | - | 3 | 351 | ||
| AK009245.1 | - | 3 | 376 | |||
| AK151108.1 | - | 3 | 328 | |||
| AK151400.1 | - | 3 | 328 | |||
| AK152129.1 | - | 3 | 441 | |||
| AK159961.1 | - | 3 | 330 | |||
| AK167830.1 | - | 3 | 317 | |||
| AK179135.1 | - | 1 | 63 | |||
| BC016519.1 | - | 3 | 366 | 3495638 | ||
| BC090403.1 | - | 3 | 411 | 5293337 | ||

Rtca
Genomic Map