siRNA Details
| Gene Symbol: Tsfm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025537.3 | NP_079813.1 | 6 | 647 | ||
| GenBank mRNA | AJ277891.1 | - | 5 | 484 | ||
| AK009898.1 | - | 6 | 668 | |||
| AK012240.1 | - | 6 | 638 | |||
| AK018591.1 | - | 6 | 636 | |||
| AK169164.1 | - | 6 | 638 | |||
| AK171656.1 | - | 6 | 644 | |||
| AK172112.1 | - | 6 | 644 | |||
| AK180094.1 | - | 2 | 161 | |||
| BC034286.1 | - | 6 | 626 | |||
| BC051508.1 | - | 3 | 228 | |||
| BC057904.1 | - | 6 | 647 | 5012380 | ||

Tsfm
Genomic Map