siRNA Details
| Gene Symbol: Tsfm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025537.3 | NP_079813.1 | 5, 6 | 576 | ||
| GenBank mRNA | AJ277891.1 | - | 4, 5 | 413 | ||
| AK009898.1 | - | 5, 6 | 597 | |||
| AK012240.1 | - | 5, 6 | 567 | |||
| AK018591.1 | - | 5, 6 | 565 | |||
| AK169164.1 | - | 5, 6 | 567 | |||
| AK171656.1 | - | 5, 6 | 573 | |||
| AK172112.1 | - | 5, 6 | 573 | |||
| AK180094.1 | - | 1, 2 | 90 | |||
| BC034286.1 | - | 5, 6 | 555 | |||
| BC051508.1 | - | 2, 3 | 157 | |||
| BC057904.1 | - | 5, 6 | 576 | 5012380 | ||

Tsfm
Genomic Map