siRNA Details
| Gene Symbol: Spc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003408.1 | - | 5, 6 | 507 | ||
| AK011241.1 | - | 6, 7 | 660 | |||
| AK146727.1 | - | 6, 7 | 1135 | |||
| AK151554.1 | - | 6, 7 | 666 | |||
| BC027121.1 | - | 6, 7 | 633 | 5359960 | ||
| BC033605.1 | - | 6, 7 | 882 | 5369405 | ||
| CX222514.1 | - | 6, 7 | 692 | |||

Spc25
Genomic Map