siRNA Details
| Gene Symbol: Exoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025588.2 | NP_079864.1 | 13 | 1495 | ||
| GenBank mRNA | AK016532.1 | - | 14 | 1622 | ||
| AK050068.1 | - | 13 | 1503 | |||
| AK202862.1 | - | 3 | 233 | |||
| BC031207.1 | - | 6 | 537 | |||
| BC048154.1 | - | 13 | 1495 | 6400932 | ||
| BC049102.1 | - | 13 | 1495 | 5702581 | ||

Exoc2
Genomic Map