siRNA Details
| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025599.3 | NP_079875.2 | 4 | 453 | ||
| GenBank mRNA | AK012168.1 | - | 4 | 434 | ||
| AK019979.1 | - | 4 | 429 | |||
| BC038891.1 | - | 4 | 340 | 3371329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmss1| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025599.3 | NP_079875.2 | 4 | 453 | ||
| GenBank mRNA | AK012168.1 | - | 4 | 434 | ||
| AK019979.1 | - | 4 | 429 | |||
| BC038891.1 | - | 4 | 340 | 3371329 | ||