siRNA Details
| Gene Symbol: Cul7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025611.5 | NP_079887.3 | 21 | 4270 | ||
| GenBank mRNA | AK147156.1 | - | 21 | 4028 | ||
| AK147905.1 | - | 21 | 3989 | |||
| AK170808.1 | - | 21 | 4043 | |||
| AK172882.1 | - | 20 | 3343 | |||
| BC019645.1 | - | 11 | 1558 | |||
| BC026946.1 | - | 3 | 378 | |||
| BC059865.1 | - | 17 | 2786 | |||
| BC065096.1 | - | 20 | 3757 | |||
| BC138998.1 | - | 21 | 4208 | 8862020 | ||
| BC139000.1 | - | 21 | 4208 | 8862022 | ||

Cul7
Genomic Map