siRNA Details
| Gene Symbol: Timm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025616.3 | NP_079892.1 | 6 | 447 | ||
| GenBank mRNA | AK008340.1 | - | 6 | 448 | ||
| AK008382.1 | - | 6 | 445 | |||
| AK010832.1 | - | 6 | 413 | |||
| AK012977.1 | - | 6 | 444 | |||
| AK088074.1 | - | 6 | 440 | |||
| AK167473.1 | - | 6 | 450 | |||
| AY551342.1 | - | 6 | 433 | |||
| BC010303.1 | - | 6 | 422 | 3597676 | ||
| BC037056.1 | - | 6 | 449 | |||

Timm50
Genomic Map