siRNA Details
| Gene Symbol: Timm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025616.3 | NP_079892.1 | 9, 10 | 872 | ||
| GenBank mRNA | AK008340.1 | - | 9, 10 | 873 | ||
| AK008382.1 | - | 9, 10 | 869 | |||
| AK010832.1 | - | 9, 10 | 838 | |||
| AK088074.1 | - | 9, 10 | 863 | |||
| AK167473.1 | - | 9, 10 | 875 | |||
| AY551342.1 | - | 9, 10 | 858 | |||
| BC010303.1 | - | 9, 10 | 847 | 3597676 | ||
| BC025844.1 | - | 3, 4 | 353 | |||
| BC037056.1 | - | 9, 10 | 874 | |||

Timm50
Genomic Map