siRNA Details
| Gene Symbol: Timm50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025616.3 | NP_079892.1 | 7 | 579 | ||
| GenBank mRNA | AK008340.1 | - | 7 | 580 | ||
| AK008382.1 | - | 7 | 577 | |||
| AK010832.1 | - | 7 | 545 | |||
| AK088074.1 | - | 7 | 572 | |||
| AK167473.1 | - | 7 | 582 | |||
| AK179970.1 | - | 2 | 101 | |||
| AY551342.1 | - | 7 | 565 | |||
| BC010303.1 | - | 7 | 554 | 3597676 | ||
| BC025844.1 | - | 1 | 60 | |||
| BC037056.1 | - | 7 | 581 | |||

Timm50
Genomic Map