siRNA Details
Gene Symbol: Aggf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025630.3 | NP_079906.2 | 14 | 2361 | ||
GenBank mRNA | AK009533.1 | - | 10 | 1324 | ||
AY500995.1 | - | 14 | 2334 | |||
BC027286.1 | - | 9 | 1037 | |||
BC028451.1 | - | 1 | 496 | |||
BC052410.1 | - | 14 | 2338 | 5716440 |
