siRNA Details
| Gene Symbol: Sppl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023220.2 | NP_075709.2 | 2 | 390 | ||
| GenBank mRNA | AB041547.1 | - | 2 | 390 | ||
| AK008294.1 | - | 2 | 177 | |||
| AK008320.1 | - | 2 | 178 | |||
| AK008328.1 | - | 2 | 177 | |||
| AK155011.1 | - | 2 | 191 | |||
| BC026578.1 | - | 2 | 198 | 5059823 | ||

Sppl2a
Genomic Map