siRNA Details
| Gene Symbol: Metap1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025633.3 | NP_079909.1 | 6 | 584 | ||
| GenBank mRNA | AJ414378.1 | - | 3 | 447 | ||
| AK005241.1 | - | 6 | 581 | |||
| AK010067.1 | - | 6 | 584 | |||
| AK043123.1 | - | 3 | 1370 | |||
| AK139461.1 | - | 1495 | ||||
| BC051534.1 | - | 3 | 458 | 3466745 | ||
| BC060739.1 | - | 5 | 2092 | |||

Metap1d
Genomic Map