siRNA Details
| Gene Symbol: Esf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081090.1 | NP_001074559.1 | 2 | 243 | ||
| GenBank mRNA | AK008642.1 | - | 2 | 235 | ||
| AK040644.1 | - | 2 | 236 | |||
| AK076024.1 | - | 2 | 237 | |||
| AK132230.1 | - | 1 | 165 | |||
| AK163125.1 | - | 1 | 85 | |||
| BC010672.1 | - | 2 | 212 | |||
| BC021330.1 | - | 2 | 260 | |||
| BC055464.1 | - | 1 | 144 | |||
| BC096029.1 | - | 2 | 212 | |||
| BC150893.1 | - | 2 | 209 | 9087804 | ||
| BC157979.1 | - | 2 | 209 | 9007299 | ||

Esf1
Genomic Map