siRNA Details
| Gene Symbol: Esf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081090.1 | NP_001074559.1 | 2, 3 | 773 | ||
| GenBank mRNA | AK076024.1 | - | 2, 3 | 767 | ||
| AK132230.1 | - | 1, 2 | 695 | |||
| AK163125.1 | - | 1, 2 | 616 | |||
| BC010672.1 | - | 2, 3 | 744 | |||
| BC021330.1 | - | 2, 3 | 792 | |||
| BC038028.1 | - | 1, 2 | 265 | |||
| BC055464.1 | - | 1, 2 | 675 | |||
| BC096029.1 | - | 2, 3 | 770 | |||
| BC150893.1 | - | 2, 3 | 739 | 9087804 | ||
| BC157979.1 | - | 2, 3 | 739 | 9007299 | ||

Esf1
Genomic Map