siRNA Details
| Gene Symbol: Fastk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023229.2 | NP_075718.2 | 5 | 962 | ||
| GenBank mRNA | AF168682.1 | - | 4 | 857 | ||
| AK002552.1 | - | 5 | 913 | |||
| AK003940.1 | - | 2 | 205 | |||
| AK038596.1 | - | 5 | 914 | |||
| AK052667.1 | - | 5 | 907 | |||
| AK139374.1 | - | 4 | 1681 | |||
| AK151859.1 | - | 5 | 914 | |||
| AK152369.1 | - | 5 | 963 | |||
| AK159751.1 | - | 5 | 914 | |||
| AK164317.1 | - | 920 | ||||
| AK166508.1 | - | 1503 | ||||
| AK167346.1 | - | 4 | 1674 | |||
| AK179538.1 | - | 2 | 131 | |||
| BC013547.1 | - | 5 | 879 | 4218374 | ||

Fastk
Genomic Map