siRNA Details
| Gene Symbol: Trim13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164220.1 | NP_001157692.1 | 3 | 459 | ||
| NM_023233.3 | NP_075722.1 | 3 | 397 | |||
| GenBank mRNA | AF220129.1 | - | 3 | 474 | ||
| AK013959.2 | - | 3 | 369 | |||
| AK017850.1 | - | 2 | 354 | |||
| AK162129.1 | - | 3 | 447 | |||
| BC138576.1 | - | 2 | 323 | 8861596 | ||
| BC145915.1 | - | 2 | 323 | 40131218 | ||
| BM933374.1 | - | 3 | 397 | |||

Trim13
Genomic Map