siRNA Details
| Gene Symbol: Hiatl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083901.1 | NP_001077370.1 | 12 | 1450 | ||
| NM_133680.3 | NP_598441.3 | 11 | 1547 | |||
| GenBank mRNA | AK146596.1 | - | 12 | 1443 | ||
| AK190739.1 | - | 1 | 21 | |||
| BC018395.1 | - | 4 | 335 | |||
| BC033469.1 | - | 11 | 1509 | 4951631 | ||

Hiatl1
Genomic Map