siRNA Details
| Gene Symbol: Dph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025675.4 | NP_079951.1 | 2 | 155 | ||
| GenBank mRNA | AK006965.1 | - | 2 | 113 | ||
| AK007021.1 | - | 2 | 158 | |||
| AK017595.1 | - | 2 | 111 | |||
| AK033888.1 | - | 2 | 120 | |||
| AK036021.1 | - | 2 | 192 | |||
| AK082013.1 | - | 2 | 192 | |||
| AK088160.1 | - | 2 | 157 | |||
| AK145789.1 | - | 2 | 115 | |||
| BC022995.1 | - | 2 | 180 | 5363234 | ||

Dph6
Genomic Map