siRNA Details
| Gene Symbol: Dph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025675.4 | NP_079951.1 | 9 | 851 | ||
| GenBank mRNA | AK006965.1 | - | 9 | 809 | ||
| AK007021.1 | - | 9 | 854 | |||
| AK017595.1 | - | 9 | 807 | |||
| AK088160.1 | - | 9 | 853 | |||
| AK145789.1 | - | 9 | 854 | |||
| BC022995.1 | - | 8 | 781 | 5363234 | ||

Dph6
Genomic Map