siRNA Details
Gene Symbol: Eef1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285429.1 | NP_001272358.1 | 3 | 272 | ||
NM_001285430.1 | NP_001272359.1 | 3 | 272 | |||
NM_023240.3 | NP_075729.2 | 3 | 275 | |||
NM_029663.2 | NP_083939.1 | 5 | 1420 | |||
GenBank mRNA | AF304351.1 | - | 3 | 238 | ||
AK010347.1 | - | 4 | 250 | |||
AK017796.1 | - | 3 | 238 | |||
AK044934.1 | - | 3 | 227 | |||
AK054335.1 | - | 3 | 229 | |||
BC079855.1 | - | 5 | 1366 | 6403006 | ||
BQ177630.1 | - | 3 | 242 | |||
BY035199.1 | - | 3 | 277 | |||
BY104511.1 | - | 3 | 263 |
