siRNA Details
| Gene Symbol: Acp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019800.4 | NP_062774.2 | 6, 7 | 1149 | ||
| GenBank mRNA | AB030038.1 | - | 6, 7 | 1142 | ||
| AF216223.1 | - | 6, 7 | 626 | |||
| AK002783.1 | - | 6, 7 | 1063 | |||
| AK077729.1 | - | 6, 7 | 1137 | |||
| AK160271.1 | - | 6, 7 | 1068 | |||
| AK165356.1 | - | 6, 7 | 1150 | |||
| AK171757.1 | - | 6, 7 | 1064 | |||
| BC047276.1 | - | 6, 7 | 889 | 6487551 | ||

Acp6
Genomic Map