siRNA Details
| Gene Symbol: Sorcs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025696.3 | NP_079972.1 | 26 | 3669 | ||
| GenBank mRNA | AF276314.1 | - | 26 | 3689 | ||
| AK158805.1 | - | 26 | 3536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sorcs3| Gene Symbol: Sorcs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025696.3 | NP_079972.1 | 26 | 3669 | ||
| GenBank mRNA | AF276314.1 | - | 26 | 3689 | ||
| AK158805.1 | - | 26 | 3536 | |||