siRNA Details
| Gene Symbol: Tmem186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025708.4 | NP_079984.2 | 2 | 567 | ||
| GenBank mRNA | AK009236.1 | - | 2 | 404 | ||
| AK013147.1 | - | 2 | 396 | |||
| AK014478.1 | - | 2 | 568 | |||
| AK033686.1 | - | 2 | 405 | |||
| AK088656.1 | - | 2 | 577 | |||
| AK159727.1 | - | 2 | 553 | |||
| AK159782.1 | - | 2 | 553 | |||
| BC027261.1 | - | 2 | 397 | 2649343 | ||
| BC058268.1 | - | 1 | 1629 | |||

Tmem186
Genomic Map