siRNA Details
| Gene Symbol: Aspn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172481.1 | NP_001165952.1 | 4 | 677 | ||
| NM_025711.3 | NP_079987.2 | 4 | 694 | |||
| GenBank mRNA | AF316825.1 | - | 3 | 414 | ||
| AK014504.1 | - | 4 | 650 | |||
| AK085920.1 | - | 4 | 649 | |||
| BC034888.1 | - | 4 | 596 | 1365428 | ||
| BC145904.1 | - | 4 | 592 | 40131207 | ||
| BC145930.1 | - | 4 | 575 | 40131233 | ||

Aspn
Genomic Map