siRNA Details
| Gene Symbol: Nkapl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025719.3 | NP_079995.2 | 1 | 1091 | ||
| GenBank mRNA | AK014812.1 | - | 1 | 1078 | ||
| AK160424.1 | - | 1 | 893 | |||
| BC099550.1 | - | 1 | 1091 | 30918538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkapl| Gene Symbol: Nkapl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025719.3 | NP_079995.2 | 1 | 1091 | ||
| GenBank mRNA | AK014812.1 | - | 1 | 1078 | ||
| AK160424.1 | - | 1 | 893 | |||
| BC099550.1 | - | 1 | 1091 | 30918538 | ||