siRNA Details
| Gene Symbol: Sbds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023248.1 | NP_075737.1 | 3 | 514 | ||
| GenBank mRNA | AK005079.1 | - | 3 | 491 | ||
| AK014648.1 | - | 3 | 519 | |||
| AK019228.1 | - | 3 | 501 | |||
| AK168847.1 | - | 3 | 501 | |||
| BC003849.1 | - | 3 | 514 | 3599733 | ||

Sbds
Genomic Map