siRNA Details
Gene Symbol: 4921522P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007249.1 | - | 2 | 707 | ||
AK014948.1 | - | 2 | 707 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4921522P10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007249.1 | - | 2 | 707 | ||
AK014948.1 | - | 2 | 707 |