siRNA Details
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025730.3 | NP_080006.3 | 43 | 6374 | ||
GenBank mRNA | AK014886.1 | - | 4 | 433 | ||
AK014938.1 | - | 4 | 446 | |||
AY792512.1 | - | 43 | 6317 | |||
BC034074.1 | - | 9 | 1257 |
