siRNA Details
| Gene Symbol: Kcng4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025734.2 | NP_080010.2 | 3 | 1804 | ||
| GenBank mRNA | AF450109.1 | - | 3 | 1470 | ||
| AK015001.1 | - | 1 | 271 | |||
| AK015820.1 | - | 1 | 82 | |||
| BC043936.1 | - | 3 | 1804 | 4504272 | ||

Kcng4
Genomic Map