siRNA Details
| Gene Symbol: Zfp474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025749.3 | NP_080025.3 | 2 | 510 | ||
| GenBank mRNA | AK016749.1 | - | 2 | 511 | ||
| AY350709.1 | - | 4 | 692 | |||
| BC055099.1 | - | 2 | 506 | 6704066 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp474| Gene Symbol: Zfp474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025749.3 | NP_080025.3 | 2 | 510 | ||
| GenBank mRNA | AK016749.1 | - | 2 | 511 | ||
| AY350709.1 | - | 4 | 692 | |||
| BC055099.1 | - | 2 | 506 | 6704066 | ||