siRNA Details
| Gene Symbol: Zfp474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025749.3 | NP_080025.3 | 2 | 960 | ||
| GenBank mRNA | AK007266.1 | - | 1 | 12 | ||
| AK016749.1 | - | 2 | 961 | |||
| AY350709.1 | - | 4 | 1142 | |||
| BC055099.1 | - | 2 | 956 | 6704066 | ||

Zfp474
Genomic Map