siRNA Details
Gene Symbol: Gid4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025757.4 | NP_080033.3 | 4 | 955 | ||
GenBank mRNA | AJ404329.1 | - | 4 | 443 | ||
AK012042.1 | - | 4 | 783 | |||
AK017120.1 | - | 955 | ||||
AK039435.1 | - | 4 | 839 | |||
AK077759.1 | - | 4 | 522 | |||
AK142753.1 | - | 4 | 757 | |||
AK164644.1 | - | 4 | 822 | |||
BC044901.1 | - | 4 | 657 | 3660276 | ||
BC046821.1 | - | 4 | 769 | 6400140 | ||
BC052418.1 | - | 4 | 797 | 6401069 |
