siRNA Details
| Gene Symbol: Cntnap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004357.2 | NP_001004357.2 | 24 | 4087 | ||
| NM_025771.3 | NP_080047.1 | 4 | 534 | |||
| GenBank mRNA | AK013139.1 | - | 4 | 502 | ||
| AK017341.1 | - | 4 | 529 | |||
| AK129232.1 | - | 24 | 4060 | |||
| BC029826.1 | - | 4 | 483 | |||
| BC064467.1 | - | 19 | 2978 | |||
| BC103799.1 | - | 4 | 441 | |||
| BC132074.1 | - | 3 | 346 | |||
| BC132076.1 | - | 3 | 346 | |||

Cntnap2
Genomic Map