siRNA Details
| Gene Symbol: Bcs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012324.1 | - | 2 | 498 | ||
| AK078925.1 | - | 2 | 510 | |||
| AK079385.1 | - | 2 | 506 | |||
| BC019781.1 | - | 2 | 398 | 4036047 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcs1l| Gene Symbol: Bcs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012324.1 | - | 2 | 498 | ||
| AK078925.1 | - | 2 | 510 | |||
| AK079385.1 | - | 2 | 506 | |||
| BC019781.1 | - | 2 | 398 | 4036047 | ||