siRNA Details
| Gene Symbol: 0610009O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024179.5 | NP_077141.2 | 12 | 1622 | ||
| GenBank mRNA | AK002437.1 | - | 12 | 1612 | ||
| AK009400.1 | - | 5 | 634 | |||
| AK141675.1 | - | 1585 | ||||
| AK171837.1 | - | 12 | 1613 | |||
| AK172900.1 | - | 12 | 1547 | |||

0610009O20Rik
Genomic Map