siRNA Details
| Gene Symbol: Etfdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025794.2 | NP_080070.2 | 13 | 1943 | ||
| GenBank mRNA | AK002483.1 | - | 13 | 1896 | ||
| AK075673.1 | - | 13 | 1894 | |||
| AK141684.1 | - | 13 | 1897 | |||
| AK152624.1 | - | 13 | 1912 | |||
| AK169686.1 | - | 13 | 1848 | |||
| BC012522.1 | - | 13 | 1892 | 2648103 | ||
| BC057670.1 | - | 13 | 1910 | 2810976 | ||

Etfdh
Genomic Map