siRNA Details
| Gene Symbol: Hmg20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025812.2 | NP_080088.1 | 9 | 1117 | ||
| GenBank mRNA | AK004596.1 | - | 9 | 1137 | ||
| AK017716.1 | - | 9 | 1320 | |||
| AK039222.1 | - | 8 | 2131 | |||
| AK129010.1 | - | 9 | 1319 | |||
| AK134213.1 | - | 9 | 1289 | |||
| BC013804.1 | - | 9 | 1117 | 3602239 | ||
| DQ182735.1 | - | 9 | 1117 | |||

Hmg20a
Genomic Map