siRNA Details
| Gene Symbol: Mfsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025813.3 | NP_080089.1 | 4, 5 | 410 | ||
| GenBank mRNA | AK004586.1 | - | 4, 5 | 411 | ||
| AK032688.1 | - | 4, 5 | 379 | |||
| AK075859.1 | - | 4, 5 | 440 | |||
| AK135235.1 | - | 4, 5 | 411 | |||
| AK136108.1 | - | 4, 5 | 402 | |||
| AK152444.1 | - | 4, 5 | 432 | |||
| AK159554.1 | - | 4, 5 | 438 | |||
| AK218750.1 | - | 2, 3 | 122 | |||

Mfsd1
Genomic Map