siRNA Details
| Gene Symbol: Appbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025825.3 | NP_080101.1 | 3 | 572 | ||
| GenBank mRNA | AK004888.1 | - | 3 | 583 | ||
| AK005440.1 | - | 3 | 278 | |||
| AK122222.1 | - | 179 | ||||
| AK133853.1 | - | 3 | 564 | |||
| AK182121.1 | - | 2 | 138 | |||
| BC018442.1 | - | 3 | 550 | 3157290 | ||
| BC019152.1 | - | 3 | 573 | 5065139 | ||

Appbp2
Genomic Map